missing translation for 'onlineSavingsMsg'
Learn More

ROBU Rubber Sleeves

Rubber Sleeves for use with a crucible.

Brand:  ROBU 20702

 View more versions of this product

Product Code. 16843026

  • 130.00 SEK / Pack of 5
Estimated Shipment: 31-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Seals filter crucible and corresponding adapter
Specifications

Specifications

For 15 mL Filter Crucibles
Natural Rubber
33 mm
25 mm
5/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers