missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBU Rubber Sleeves
Rubber Sleeves for use with a crucible.
123.00 SEK - 139.00 SEK
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
16833026
|
ROBU
20701 |
26 mm |
123.00 SEK
Pack of 5 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
15327006
|
ROBU
20703 |
41 mm |
139.00 SEK
Pack of 5 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||||
16843026
|
ROBU
20702 |
33 mm |
130.00 SEK
Pack of 5 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
Description
- Seals filter crucible and corresponding adapter
Specifications
Natural Rubber | |
5/Pk. |