Learn More
Wheaton™ Scintillation Vial
Brand: Wheaton™ 227180
2455.76 SEK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Specifications
500 | |
500 Pk. |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
19Hours
17Minutes
33Seconds
26 -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
50Hours
17Minutes
33Seconds
26New Offer Save Up to 70%