Learn More
Hirschmann™ SILICOSEN™ Silicone Stoppers
Excellent for the cultivation of aerobic microorganisms. Specially processed culture plugs with continuous bubbles for preparation, filling and sterilization of culture media.
Brand:  Hirschmann™ 8905828
 View more versions of this product
Product Code. 10651604
  • 2345.00 SEK / Pack of 10
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Description
  • Uniform pore structure
  • Good air permeability
  • Chemically and temperature resistant
  • Liquid repellent and reusable
Specifications
Specifications
SILICOSEN, S28
For The Preparation And Sterilization Of Culture Media
18 mm
Green
28 mm
Silicone Stopper
17-26 mm
27 mm
10/Pack
17 mm
Silicone