missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ SILICOSEN™ Silicone Stoppers
Excellent for the cultivation of aerobic microorganisms. Specially processed culture plugs with continuous bubbles for preparation, filling and sterilization of culture media.
708.00 SEK - 3540.00 SEK
Specifications
Material | Silicone |
---|---|
Product Type | Silicone Stopper |
For Use With (Application) | For The Preparation And Sterilization Of Culture Media |
Quantity | 10/Pack |
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
10670644
|
Hirschmann™
8905612 |
9-11 mm |
708.00 SEK
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10716843
|
Hirschmann™
8905720 |
15-20 mm |
1150.00 SEK
Pack of 10 |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
10651604
|
Hirschmann™
8905828 |
17-26 mm |
2345.00 SEK
Pack of 10 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
10706843
|
Hirschmann™
8905622 |
18-21 mm |
1220.00 SEK
Pack of 10 |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
10047512
|
Hirschmann™
8905840 |
28-38 mm |
3540.00 SEK
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15514859
|
Hirschmann™
8905765 |
42-52 mm |
3425.00 SEK
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Uniform pore structure
- Good air permeability
- Chemically and temperature resistant
- Liquid repellent and reusable
Specifications
Silicone | |
Silicone Stopper | |
For The Preparation And Sterilization Of Culture Media | |
10/Pack |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title