Learn More
Hirschmann™ SILICOSEN™ Silicone Stoppers
Excellent for the cultivation of aerobic microorganisms. Specially processed culture plugs with continuous bubbles for preparation, filling and sterilization of culture media.
Brand: Hirschmann™ 8905765
This item is not returnable.
View return policy
Description
- Uniform pore structure
- Good air permeability
- Chemically and temperature resistant
- Liquid repellent and reusable
Specifications
SILICOSEN, C65 | |
42-52 mm | |
29 mm | |
Natural | |
57 mm | |
Silicone Stopper |
42 mm | |
For The Preparation And Sterilization Of Culture Media | |
10/Pack | |
65 mm | |
Silicone |