missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21764-1-AP-150UL
This item is not returnable.
View return policy
Description
Located between the protocadherin beta and gamma gene clusters on chromosome 5, this intronless gene encodes a protein that is highly similar to an ornithine transporter localized in the mitochondrial inner membrane. The encoded protein most likely plays a role in metabolism as a mitochondrial transport protein.Specifications
SLC25A2 | |
Polyclonal | |
Unconjugated | |
SLC25A2 | |
ORC2, ORNT2, SLC25A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
291640, 83884, 83885 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BXI2 | |
Slc25a2 | |
SLC25A2 Fusion Protein Ag16360 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |