missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SLC25A2 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16811365
|
Proteintech
21764-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16801365
|
Proteintech
21764-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Located between the protocadherin beta and gamma gene clusters on chromosome 5, this intronless gene encodes a protein that is highly similar to an ornithine transporter localized in the mitochondrial inner membrane. The encoded protein most likely plays a role in metabolism as a mitochondrial transport protein.Specifications
SLC25A2 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q9BXI2 | |
291640, 83884, 83885 | |
SLC25A2 Fusion Protein Ag16360 | |
Primary | |
-20°C | |
SLC25A2 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ORC2, ORNT2, SLC25A2 | |
Slc25a2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |