Learn More
ICU Medical Jelco™ Saf-T Holder™ Blood Culture Holders and Transfer Devices
Saf-T Holder™ devices come with the back-end needle and multi-sample Luer permanently attached to the holder to minimize potential exposures and holder re-use. Available separately or with Saf-T Wing™ Blood Collection & Infusion Sets, Saf-T Holder™ devices also provide easy needless transfer after a syringe draw.
Brand: ICU Medical 96004
Description
- Eliminate potential exposures and holder reuse
- Provide safe needleless transfer after a syringe draw
Specifications
Multi-sample Luer permanently attached to the holder, provide easy needless transfer after a syringe draw | |
Saf-T Holder™ Blood Culture Device, with male luer |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.