Learn More
ICU Medical Jelco™ Saf-T Holder™ Blood Culture Holders and Transfer Devices
Saf-T Holder™ devices come with the back-end needle and multi-sample Luer permanently attached to the holder to minimize potential exposures and holder re-use. Available separately or with Saf-T Wing™ Blood Collection & Infusion Sets, Saf-T Holder™ devices also provide easy needless transfer after a syringe draw.
1041.84 SEK
Specifications
For Use With (Application) Multi-sample Luer permanently attached to the holder, provide easy needless transfer after a syringe draw
View More Specs
Products 1
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
15785068
Encompass
View Documents
ICU Medical
96004
Saf-T Holder™ Blood Culture Device, with male luer
1041.84 SEK
Pack of 50
View alternative products
Save Up to 
»
Description
Description
  • Eliminate potential exposures and holder reuse
  • Provide safe needleless transfer after a syringe draw
Specifications
Specifications
Multi-sample Luer permanently attached to the holder, provide easy needless transfer after a syringe draw