Learn More
SMYD4 Mouse anti-Human, Unconjugated, Clone: 2F4B7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66874-1-IG-150UL
 View more versions of this product
Product Code. 16879135
  • 4721.67 SEK / 150µL
Explore more special offers
Description
Description
The SMYD4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMYD4. This antibody recognizes human antigen. The SMYD4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SMYD4
Monoclonal
920 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
2F4B7
Unconjugated
150 μL
114826
IgG1