missing translation for 'onlineSavingsMsg'
Learn More

SMYD4 Mouse anti-Human, Unconjugated, Clone: 2F4B7, Proteintech

Mouse Monoclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen SMYD4
Clone 2F4B7
Concentration 920 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16889135
View Documents
Proteintech
66874-1-IG-20UL
20 μL
1656.73 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16879135
View Documents
Proteintech
66874-1-IG-150UL
150 μL
4721.67 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SMYD4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMYD4. This antibody recognizes human antigen. The SMYD4 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

SMYD4
920 μg/mL
Monoclonal
Mouse
114826
Primary
2F4B7
Western Blot, ELISA
Unconjugated
Human
IgG1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.