Learn More
Sodium 4-hydroxybenzenesulfonate dihydrate, 97%, Thermo Scientific Chemicals
395.00 SEK - 4100.00 SEK
Chemical Identifiers
CAS | 10580-19-5 |
---|---|
Molecular Formula | C6H5O4S |
Molecular Weight (g/mol) | 173.16 |
MDL Number | MFCD00044734,MFCD00150724 |
InChI Key | FEPBITJSIHRMRT-UHFFFAOYSA-M |
Synonym | sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate |
PubChem CID | 23666329 |
IUPAC Name | sodium;4-hydroxybenzenesulfonate;dihydrate |
SMILES | OC1=CC=C(C=C1)S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11481950
|
Thermo Scientific Alfa Aesar
A14808.22 |
100 g |
395.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11491950
|
Thermo Scientific Alfa Aesar
A14808.36 |
500 g |
1160.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15437515
|
Thermo Scientific Alfa Aesar
A14808.0E |
2500 g |
4100.00 SEK
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10580-19-5 | |
173.16 | |
FEPBITJSIHRMRT-UHFFFAOYSA-M | |
23666329 | |
OC1=CC=C(C=C1)S([O-])(=O)=O |
C6H5O4S | |
MFCD00044734,MFCD00150724 | |
sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate | |
sodium;4-hydroxybenzenesulfonate;dihydrate |
Specifications
10580-19-5 | |
Odorless | |
MFCD00044734,MFCD00150724 | |
sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate | |
OC1=CC=C(C=C1)S([O-])(=O)=O | |
173.16 | |
232.20 (196.17 Anhydrous) | |
Sodium 4-hydroxybenzenesulfonate dihydrate |
>300°C | |
C6H5O4S | |
4040891 | |
FEPBITJSIHRMRT-UHFFFAOYSA-M | |
sodium;4-hydroxybenzenesulfonate;dihydrate | |
23666329 | |
97% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 212-550-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium 4-hydroxybenzenesulfonate dihydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sodium 4-hydroxybenzenesulfonate dihydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.