missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Metabisulfite, Reagent grade, 97%, Honeywell Burdick & Jackson™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Melting Point | >150°C |
Color | White |
pH | 3.5 to 5 (at 50g/L 20°C) |
Quantity | 2.5 kg |
Linear Formula | Na2S2O5 |
UN Number | NONH for all modes of transport |
Solubility Information | Soluble in water, 650g/L at 20°C |
Formula Weight | 190.11g/mol |
Percent Purity | 97% |
Grade | Reagent |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Metabisulfite, Reagent grade, 97%, Honeywell Burdick & Jackson™ >
Spot an opportunity for improvement?Share a Content Correction