missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metabisulfite, Reagent grade, 97%, Honeywell Burdick & Jackson™

Reagent Grade, 97%

369.77 SEK - 662.41 SEK

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15671530
SDS View Documents
Honeywell Research Chemicals
161519-500G
500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15661530
SDS View Documents
Honeywell Research Chemicals
161519-2.5KG
2.5 kg Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P280-P301 + P312 + P330-P305 + P351 + P338 + P310

H302-H318

missing translation for 'einecsNumber' : 231-673-0

missing translation for 'rtecsNumber' : UX8225000

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Metabisulfite, Reagent grade, 97%, Honeywell Burdick & Jackson™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.