missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tetrathionate Dihydrate, purum p.a, ≥98.0% (T), Honeywell™
Purum p.a.,≥98.0% (T)
2075.85 SEK - 6301.77 SEK
Chemical Identifiers
CAS | 13721-29-4 |
---|---|
Molecular Formula | H4Na2O8S4 |
Molecular Weight (g/mol) | 306.24 |
MDL Number | MFCD00150868 |
InChI Key | HAEPBEMBOAIUPN-UHFFFAOYSA-L |
Synonym | sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2 |
PubChem CID | 203055 |
IUPAC Name | disodium dihydrate (sulfonatodisulfanyl)sulfonate |
SMILES | O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
13721-29-4 | |
306.24 | |
HAEPBEMBOAIUPN-UHFFFAOYSA-L | |
203055 | |
O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O |
H4Na2O8S4 | |
MFCD00150868 | |
sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2 | |
disodium dihydrate (sulfonatodisulfanyl)sulfonate |
Specifications
13721-29-4 | |
NaO3SS2SO3Na · 2H2O | |
NONH for all modes of transport | |
HAEPBEMBOAIUPN-UHFFFAOYSA-L | |
disodium dihydrate (sulfonatodisulfanyl)sulfonate | |
203055 | |
≥98.0% (T) |
H4Na2O8S4 | |
MFCD00150868 | |
sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2 | |
O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O | |
306.24 | |
306.27g/mol | |
Sodium tetrathionate dihydrate |
Safety and Handling
P261-P305 + P351 + P338
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Tetrathionate Dihydrate, purum p.a, ≥98.0% (T), Honeywell™