Learn More
Spectrum Labs™ Spectra/Mesh™ Woven Nylon Filter Discs
Hydrophilic nylon filters with high strength for use in laboratory, industrial, or pharmaceutical applications.
Brand:  Spectrum Labs™ 148146
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15300962
  • 1910.00 SEK / Pack of 10
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
Description
Description
  • Hydrophilic
  • Good resistance against corrosives
  • Resistant against pH 3 to 10
  • Sterilize via irradiation
Specifications
Specifications
Nylon
Laboratory, Industrial, Pharmaceutical
125 μm
200 μm
55%
47 mm