Learn More
Spectrum Labs™ Spectra/Mesh™ Woven Nylon Filter Discs
Hydrophilic nylon filters with high strength for use in laboratory, industrial, or pharmaceutical applications.
Brand: Spectrum Labs™ 145807
Additional Details : Weight : 0.05000kg
Description
- Hydrophilic
- Good resistance against corrosives
- Resistant against pH 3 to 10
- Sterilize via irradiation
Specifications
Nylon | |
Laboratory, Industrial, Pharmaceutical | |
60 μm |
41 μm | |
33% | |
55 mm |