Learn More
Nabertherm™ Split Type Vertical Tube Furnace RSV 1100 Series with P570 Controller
The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes.
Brand:  Nabertherm™ RI252V2AN3
Additional Details : Weight : 36.00000kg
 View more versions of this product
Product Code. 17388119
  • 91829.97 SEK / Each
Estimated Shipment: 28-10-2024
to see stock.
Add to basket
Description
Description
  • Maximum Temperature: 1100°C
  • Dual shell housing made of sheets of textured stainless steel
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Type N thermocouple
  • Frame for vertical operation
  • Split-type design for simple insertion of the working tube
  • Working tube made of material C 530 includes fiber plugs for operation under air in scope of delivery
  • Heating elements on support tubes provide for free radiation
  • RSV: switchgear and control unit separate from furnace in own wall or standing cabinet
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications
Specifications
P570
590 x 545 x 1225 mm
50 mm
Up to 1100°C
250 mm
36 kg