Learn More
Nabertherm™ Split Type Vertical Tube Furnace RSV 1100 Series with P570 Controller
The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes.
91829.97 SEK - 164607.59 SEK
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
17388119
|
Nabertherm™
RI252V2AN3 |
50 mm | 250 mm |
91829.97 SEK
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
17328129
|
Nabertherm™
RI322V2AN3 |
120 mm | 500 mm |
110527.31 SEK
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
17368129
|
Nabertherm™
RI382V2AN3 |
80 mm | 500 mm |
107687.21 SEK
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
17308139
|
Nabertherm™
RI422V2AN3 |
120 mm | 750 mm |
135614.88 SEK
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
17348139
|
Nabertherm™
RI472V2AN3 |
170 mm | 750 mm |
149933.73 SEK
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
17388139
|
Nabertherm™
RI482V2AN3 |
80 mm | 750 mm |
130289.69 SEK
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
17328149
|
Nabertherm™
RI522V2AN3 |
120 mm | 1000 mm |
144845.21 SEK
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
17368149
|
Nabertherm™
RI572V2AN3 |
170 mm | 1000 mm |
164607.59 SEK
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
Description
- Maximum Temperature: 1100°C
- Dual shell housing made of sheets of textured stainless steel
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Type N thermocouple
- Frame for vertical operation
- Split-type design for simple insertion of the working tube
- Working tube made of material C 530 includes fiber plugs for operation under air in scope of delivery
- Heating elements on support tubes provide for free radiation
- RSV: switchgear and control unit separate from furnace in own wall or standing cabinet
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications
P570 | |
Up to 1100°C |