Learn More
Ohaus™ Stainless Steel Weight, OIML F1, OH
Single body design weight adjusted to meet or exceed 0IML F1 tolerances
Brand:  Ohaus™ 11117042
 View more versions of this product
Product Code. 15683384
  • 1140.00 SEK / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Stainless steel construction
  • Include a weight case
  • Single body design without adjusting cavity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Weight, 200g, OIML F1, OH
Stainless Steel
Meets or exceeds OIML F1
OIML F1 Weights
200 g