Learn More
Systec™ Stainless Steel Wire Mesh Basket
Use these stackable mesh baskets with Systec™ Autoclaves. Systec Wire Mesh Baskets are made of stainless steel and cleanly welded.
Brand: Systec™ 7225
Additional Details : Weight : 2.16900kg
Description
- Benchtop autoclaves are fitted with adjustable leveling screws to ensure stability
- Fully utilize the available space in the chamber, especially when sterilizing small items
- Entire shelving system or individual trays can be removed
Specifications
885 x 240 mm | |
Wire Mesh Basket | |
1 Ea. |
Systec V-Series Autoclave models V-65 & V-95 | |
Stainless Steel | |
885 H x 240 mm dia. |