Learn More
Systec™ Stainless Steel Wire Mesh Basket
Use these stackable mesh baskets with Systec™ Autoclaves. Systec Wire Mesh Baskets are made of stainless steel and cleanly welded.
Brand:  Systec™ 7210
 View more versions of this product
Product Code. 10601534
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Benchtop autoclaves are fitted with adjustable leveling screws to ensure stability
  • Fully utilize the available space in the chamber, especially when sterilizing small items
  • Entire shelving system or individual trays can be removed
Specifications
Specifications
385 x 365 mm
Wire Mesh Basket
1 Ea.
Systec V-Series Autoclave V-65 to V95
Stainless Steel
385 H x 365 mm dia.