Learn More
Systec™ Stainless Steel Wire Mesh Basket
Use these stackable mesh baskets with Systec™ Autoclaves. Systec Wire Mesh Baskets are made of stainless steel and cleanly welded.
Brand:  Systec™ 1655
 View more versions of this product
Product Code. 11345814
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Benchtop autoclaves are fitted with adjustable leveling screws to ensure stability
  • Fully utilize the available space in the chamber, especially when sterilizing small items
  • Entire shelving system or individual trays can be removed
Specifications
Specifications
330 x 290 mm
Wire Mesh Basket
1 Ea.
Systec V-Series Autoclave models V-40 & V-55
Stainless Steel
330 H x 290 mm dia.