missing translation for 'onlineSavingsMsg'
Learn More

Systec™ Stainless Steel Wire Mesh Basket

Use these stackable mesh baskets with Systec™ Autoclaves. Systec Wire Mesh Baskets are made of stainless steel and cleanly welded.

Brand:  Systec™ 1553

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12012979

  • POA / Each
Estimated Shipment: 02-10-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • Benchtop autoclaves are fitted with adjustable leveling screws to ensure stability
  • Fully utilize the available space in the chamber, especially when sterilizing small items
  • Entire shelving system or individual trays can be removed
Specifications

Specifications

Systec D-Series Benchtop Autoclave model D45
Stainless Steel
500 L x 240 W x 220 mm H
Wire Mesh Basket
1 Ea.
500 x 240 x 220 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers