Learn More
Systec™ Stainless Steel Wire Mesh Basket
Use these stackable mesh baskets with Systec™ Autoclaves. Systec Wire Mesh Baskets are made of stainless steel and cleanly welded.
Brand: Systec™ 6072
Additional Details : Weight : 1.00000kg
Description
- Benchtop autoclaves are fitted with adjustable leveling screws to ensure stability
- Fully utilize the available space in the chamber, especially when sterilizing small items
- Entire shelving system or individual trays can be removed
Specifications
Systec D-Series Benchtop Autoclave model D-100, H-100, D-200 and H-200 | |
Stainless Steel | |
495 L x 318 W x 219 mm H |
Wire Mesh Basket | |
1 Ea. | |
495 x 318 x 219 mm |