Learn More
Systec™ Stainless Steel Wire Mesh Basket
Use these stackable mesh baskets with Systec™ Autoclaves. Systec Wire Mesh Baskets are made of stainless steel and cleanly welded.
Brand:  Systec™ 7076
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12195481
  • POA / Each
Estimated Shipment: 02-10-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Benchtop autoclaves are fitted with adjustable leveling screws to ensure stability
  • Fully utilize the available space in the chamber, especially when sterilizing small items
  • Entire shelving system or individual trays can be removed
Specifications
Specifications
Systec D-Series Benchtop Autoclave model D-150 and H-150
Stainless Steel
750 L x 370 W x 150 mm H
Wire Mesh Basket
1 Ea.
750 x 370 x 150 mm