missing translation for 'onlineSavingsMsg'
Learn More

Stat1 (pY701) Mouse, Alexa Fluor 647, Clone: 4A, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 562070

 View more versions of this product

Product Code. 15831619

  • 11165.00 SEK / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

STAT ( S ignal t ransducer and activators of nterferon- S timulated G ene F actor 3 ) complex, which is the primary transcription activator induced by the binding of the interferon to a specific cell-surface receptor. Stat1 has two alternatively spliced isoforms, 91-kDa Stat1α and 84-kDa Stat1β; Stat1α has 38 additional C-terminal amino acids. In response to the binding of IFNα, IFNγ, EGF, PDGF, or CSF-1 to their respective receptors, the Stat1 subunits become tyrosine-phosphorylated at Y701, and the complex is translocated to the nucleus. This results in the formation of an active complex that includes the DNA-binding p48 subunit. This complex is responsible for modulating the transcription of the interferon-stimulated genes (ISGs).

The 4a monoclonal antibody recognizes the phosphorylated Y701 in Stat1 α.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stat1 (pY701)
Monoclonal
Alexa Fluor 647
Mouse
Affinity Purified
RUO
Human, Mouse
IgG2a
Flow Cytometry
4a
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Phosphorylated Human Stat1 Peptide
250 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.