Learn More
Stat1 (pY701) Mouse, Alexa Fluor 647, Clone: 4A, BD
Mouse Monoclonal Antibody
3790.00 SEK - 11165.00 SEK
Specifications
Antigen | Stat1 (pY701) |
---|---|
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Host Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15836239
|
BD Biosciences
612597 |
50 Tests |
3790.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15831619
|
BD Biosciences
562070 |
250 Tests |
11165.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Stat (Signal transducer and activators of transcription) proteins mediate the biological activity of cytokines, including interleukins, interferons, erythropoietin, and growth factors. Ligand-receptor interaction activates constitutively-associated JAK family kinases as well as subsequent recruitment and activation of Stat proteins by tyrosine phosphorylation. Active Stat proteins then move to the nucleus to promote transcription of cytokine-inducible genes. Seven Stat proteins have been cloned, each of which is differentially expressed and/or activated in a cytokine-specific and cell type-specific manner. Stat1 and Stat2 are components of the ISGF3 (Interferon-Stimulated Gene Factor 3) complex, the primary transcription activator induced by interferon binding to a specific cell-surface receptor. Stat1 has two alternatively spliced isoforms, 91-kDa Stat1α and 84-kDa Stat1β; Stat1α has 38 additional C-terminal amino acids. In response to the binding of IFNα, IFNγ, EGF, PDGF, or CSF-1 to their respective receptors, the Stat1 subunits become tyrosine-phosphorylated at Y701, and the complex translocates to the nucleus. This forms an active complex that includes the DNA-binding p48 subunit, and is responsible for modulating interferon-stimulated genes (ISGs) transciption.
The 4a monoclonal antibody recognizes the phosphorylated Y701 in Stat1α and Stat1β.
Specifications
Stat1 (pY701) | |
Monoclonal | |
Mouse | |
Human, Mouse | |
Phosphorylated Human Stat1 Peptide | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
Alexa Fluor 647 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG2a | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.