Learn More
SP Bel-Art™ Sterileware Single Slot Powder Sampler
Ideal for taking a single, targeted 50 mL sample from free-flowing powders and granules
Brand:  SP Bel-Art™ F37875-0620
 View more versions of this product
Product Code. 16826926
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • After insertion, rotate tube to open then close the collection slot, and remove to retrieve the sample
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) sampler heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
50 mL
20/Pk.
62 cm