Learn More
SP Bel-Art™ Sterileware Single Slot Powder Sampler
Ideal for taking a single, targeted 50 mL sample from free-flowing powders and granules
4210.00 SEK - 5120.00 SEK
Specifications
Capacity (Metric) 50 mL
Quantity 20/Pk.
View More Specs
Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
16826926
View Documents Promotion Details
SP Bel-Art™
F37875-0620
62 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16836926
View Documents Promotion Details
SP Bel-Art™
F37875-0970
97 cm
5120.00 SEK
Pack of 20
View alternative products
Save Up to 
»
Estimated Shipment: 13-11-2024
to see stock.
Add to basket
Description
Description
  • After insertion, rotate tube to open then close the collection slot, and remove to retrieve the sample
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) sampler heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
50 mL
20/Pk.