missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Surfactants Cell Test Kits
For detecting n-Ten nonionic surfactants.
Brand: WTW™ 252061
2031.39 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- With certificate for optimum precision
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For infrequent or very large sample throughput in the laboratory
- Sample adding, inserting, measuring and reading
- Reaction cuvette with barcode or method selection

Specifications
Chromosulfuric Acid oxidation | |
PhotoLab™ S12, PhotoLab™ Spectral photometers | |
25 | |
Photometric |
For waste water, seepage water, surface water | |
Certificates | |
0.10 - 7.50mg/L Triton X-100 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Surfactants (nonion.) Cell Test Method: photometric 0.10 - 7.50 mg/l Triton X-100 Spectroquant
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H371-May cause damage to organs.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Triton X-100
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Surfactants Cell Test Kits > Cell Test; n-Ten (Nonionic); Detection Range: 0.10 to 7.5mg/L Triton X-100
Spot an opportunity for improvement?Share a Content Correction