Learn More
Vitlab™ TA II Bottletop Dispenser
Vitlab™ TA II Bottletop Dispenser designed to meet the demanding purity standards required in trace analysis.
Brand:  Vitlab™ 1627545
11445.39 SEK valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15610935
  • 13230.00 SEK / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Description
  • Includes: TA 2 dispenser (screw thread GL 45) with adjustable variable volumes, DE-M marked, with quality certificate, telescopic filling tube, mounting tool, GL 28/S 28 (ETFE), GL 32 (ETFE), and S 40 (PTFE) bottle adapters, and instruction manual
  • Can be deployed with highly concentrated acids and bases, such as perchloric, sulphuric and nitric acid
  • With tantalum spring
  • The practical screwing mechanism and inner toothed bar enable a fast, easy and precise volume adjustment
Specifications
Specifications
With recirculation