Learn More
Vitlab™ TA II Bottletop Dispenser
Vitlab™ TA II Bottletop Dispenser designed to meet the demanding purity standards required in trace analysis.
12350.00 SEK - 13230.00 SEK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15610935
|
Vitlab™
1627545 |
With recirculation |
13230.00 SEK
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
15600935
|
Vitlab™
1627535 |
Without recirculation |
12350.00 SEK
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
Description
- Includes: TA 2 dispenser (screw thread GL 45) with adjustable variable volumes, DE-M marked, with quality certificate, telescopic filling tube, mounting tool, GL 28/S 28 (ETFE), GL 32 (ETFE), and S 40 (PTFE) bottle adapters, and instruction manual
- Can be deployed with highly concentrated acids and bases, such as perchloric, sulphuric and nitric acid
- With tantalum spring
- The practical screwing mechanism and inner toothed bar enable a fast, easy and precise volume adjustment