Learn More
Tafenoquine Succinate, MedChemExpress
MedChemExpress Tafenoquine Succinate (WR 238605 Succinate) is an 8-aminoquinoline. Tafenoquine is an anti-malarial prophylactic agent.
Brand:  Medchem Express HY-111529A
Encompass_Preferred
 View more versions of this product
Product Code. 16450797
  • 6084.19 SEK / 50mg
Explore more special offers
Description
Description
Tafenoquine Succinate (WR 238605 Succinate) is an 8-aminoquinoline. Tafenoquine is an anti-malarial prophylactic agent.
Specifications
Specifications
Tafenoquine Succinate
106635-81-8
50 mg
C28H34F3N3O7
WR 238605 (Succinate)
O=C(O)CCC(O)=O.CC(NC1=C2N=C(OC)C=C(C)C2=C(OC3=CC=CC(C(F)(F)F)=C3)C(OC)=C1)CCCN
581.58
Research
Powder
Light Nude
Research
COVID-19-immunoregulation
DMSO : 125 mg/mL (214.93 mM; Need ultrasonic)
581.58
98.42%
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Tafenoquine Succinate, MedChemExpress