Learn More
Tafenoquine Succinate, MedChemExpress
MedChemExpress Tafenoquine Succinate (WR 238605 Succinate) is an 8-aminoquinoline. Tafenoquine is an anti-malarial prophylactic agent.
Brand: Medchem Express HY-111529A
Description
Tafenoquine Succinate (WR 238605 Succinate) is an 8-aminoquinoline. Tafenoquine is an anti-malarial prophylactic agent.Specifications
Tafenoquine Succinate | |
106635-81-8 | |
50 mg | |
C28H34F3N3O7 | |
WR 238605 (Succinate) | |
O=C(O)CCC(O)=O.CC(NC1=C2N=C(OC)C=C(C)C2=C(OC3=CC=CC(C(F)(F)F)=C3)C(OC)=C1)CCCN | |
581.58 | |
Research |
Powder | |
Light Nude | |
Research | |
COVID-19-immunoregulation | |
DMSO : 125 mg/mL (214.93 mM; Need ultrasonic) | |
581.58 | |
98.42% |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only