Learn More
STEMCELL Technologies Tamoxifen
Estrogen signaling pathway modulator; Modulates selective estrogen receptor and estrogen-related receptor
Brand:  STEMCELL Technologies 72664
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 17150102
  • 14004.90 SEK / 5g
Estimated Shipment: 14-08-2024
to see stock.
Description
Description
  • Tamoxifen is a selective estrogen receptor modulator (SERM), with tissue-specific antagonistic or agonistic effects
  • Receptor activation leads to the formation of homo- and hetero-dimers, which in turn interact with accessory proteins to regulate gene transcription
  • Tamoxifen is commonly used to activate Cre-ER in transgenic conditional models
  • Used in transgenic models to induce Cre-mediated recombination in conjunction with Cre-ER, a fusion protein consisting of Cre recombinase and a mutant form of the estrogen receptor hormone-binding domain that specifically binds tamoxifen but not estrogen (Feil et al.; Zhang et al.)
  • Inhibits growth in the human breast cancer cell line, MCF-7 (Katzenellenbogen et al.)
  • Antagonist of estrogen receptor action in breast tissue and breast cancer cells (Abe et al.; Horwitz et al.)
Specifications
Specifications
10540-29-1
5 g
≥95%
trans-Tamoxifen
C26H29NO