Learn More
MP Biomedicals™ Taq Core Kit 25
Taq DNA Polymerase + dNTPs in one kit
Brand:  MP Biomedicals™ 11EPTQK253
 View more versions of this product
Product Code. 11477030
  • 6670.00 SEK / 750 units
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Taq CORE Kits contain all the reagents (presented as separate items) required for PCR:
  • MP Biomedicals high quality recombinant Taq DNA polymerase, high purity dNTPs Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2 solution.
  • dNTPs at 25mM each
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3 x 250 u