Learn More
MP Biomedicals™ Taq Core Kit 25
Taq DNA Polymerase + dNTPs in one kit
6670.00 SEK
Description
- Taq CORE Kits contain all the reagents (presented as separate items) required for PCR:
- MP Biomedicals high quality recombinant Taq DNA polymerase, high purity dNTPs Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2 solution.
- dNTPs at 25mM each