Learn More
Tegafur, MedChemExpress
MedChemExpress Tegafur (FT 207; NSC 148958) is a chemotherapeutic 5-FU prodrug used in the treatment of cancers; is a component of tegafur-uracil.
Brand: Medchem Express HY-17400
This item is not returnable.
View return policy
Description
Tegafur (FT 207Specifications
Tegafur | |
White | |
C8H9FN2O3 | |
FT 207 NSC 148958 | |
O=C1NC(C(F)=CN1C2OCCC2)=O | |
200.17 | |
Research | |
Cancer-programmed cell death |
17902-23-7 | |
Research | |
100 mg | |
DMSO : ≥ 48 mg/mL (239.80 mM) | |
200.17 | |
98.74% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.