Learn More
Tegafur, MedChemExpress
MedChemExpress Tegafur (FT 207; NSC 148958) is a chemotherapeutic 5-FU prodrug used in the treatment of cancers; is a component of tegafur-uracil.
953.65 SEK - 1907.29 SEK
Specifications
Chemical Name or Material | Tegafur |
---|---|
CAS | 17902-23-7 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C8H9FN2O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16481846
|
Medchem Express
HY-17400 |
500 mg |
1907.29 SEK
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16427286
|
Medchem Express
HY-17400 |
100 mg |
953.65 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tegafur (FT 207Specifications
Tegafur | |
White | |
C8H9FN2O3 | |
DMSO : ≥ 48 mg/mL (239.80 mM) | |
200.17 | |
98.74% | |
Solid |
17902-23-7 | |
Research | |
FT 207 NSC 148958 | |
O=C1NC(C(F)=CN1C2OCCC2)=O | |
200.17 | |
Research | |
Cancer-programmed cell death |
Safety and Handling
H301∣H311∣H331
missing translation for 'shelfLife' : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Recommended Storage : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.