Learn More
Tegafur, MedChemExpress
MedChemExpress Tegafur (FT 207; NSC 148958) is a chemotherapeutic 5-FU prodrug used in the treatment of cancers; is a component of tegafur-uracil.
Brand:  Medchem Express HY-17400
Encompass_Preferred
 View more versions of this product
Product Code. 16427286
  • 953.65 SEK / 100mg
Explore more special offers
Description
Description
Tegafur (FT 207
Specifications
Specifications
Tegafur
White
C8H9FN2O3
FT 207 NSC 148958
O=C1NC(C(F)=CN1C2OCCC2)=O
200.17
Research
Cancer-programmed cell death
17902-23-7
Research
100 mg
DMSO : ≥ 48 mg/mL (239.80 mM)
200.17
98.74%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.