missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl ethyl ether, 99%, Thermo Scientific Chemicals

1065.00 SEK - 10769.20 SEK

Chemical Identifiers

CAS 637-92-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00009225
InChI Key NUMQCACRALPSHD-UHFFFAOYSA-N
Synonym tert-butyl ethyl ether, ethyl tert-butyl ether, etbe, propane, 2-ethoxy-2-methyl, ethyl tert-butyl oxide, ether, tert-butyl ethyl, 2-methyl-2-ethoxypropane, ethyl t-butyl ether, tert-butylethylether, methyl-2-ethoxypropane
PubChem CID 12512
IUPAC Name 2-ethoxy-2-methylpropane
SMILES CCOC(C)(C)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11440018
SDS View Documents
Thermo Scientific Alfa Aesar
B22945.06
5 g
1065.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
11450018
SDS View Documents
Thermo Scientific Alfa Aesar
B22945.14
25 g
3345.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
11460018
SDS View Documents
Thermo Scientific Alfa Aesar
B22945.22
100 g
10769.20 SEK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.

Solubility
Miscible with alcohol, ethyl ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, neoprene, n-butyl and urethane. It may react with air to form peroxides.
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl ethyl ether, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.