Learn More
Texwipe™ AlphaMop™ Isolator Cleaning Tool
Excellent for cleaning isolators, laminar flow hoods, biosafety cabinets, and glove boxes
Brand: Texwipe™ TX7101
1748.00 SEK valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Includes: Two handles (18 and 24 in. long), one polyester foam pad, and six TechniCloth™ polyester mop covers. Mop with telescoping handle includes six polyester mop covers.
Description
- Low-profile mop with rounded corners and swivel head to reach difficult-to-access areas
- Made from autoclavable Celcon™ plastic
- Replaceable foam pad ensures that mop cover conforms to surfaces being cleaned
Specifications
Mop | |
Autoclavable | |
Plastic |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
New Offer Save Up to 92%
Product Content Correction
Product Title
Texwipe™ AlphaMop™ Isolator Cleaning Tool > Cleaning Tool
Spot an opportunity for improvement?Share a Content Correction