Learn More
Thermo Scientific™ 24-Well Semi-Skirted PCR Plate
24-well semi-skirted plate for PCR and qPCR applications.
Brand:  Thermo Scientific™ AB-0624/R
 View more versions of this product
Product Code. 10563795
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The 24-Well Semi-Skirted PCR Plate is pre-cut into 24-well segments. Its convenient format allows for lower sample numbers. The semi skirt adds rigidity and allots space for labeling or barcoding. Available in various colors and compatible with standard multichannel pipettes. Sealing is possible with adhesive films and foils and all cap strips.
The Thermo Scientific 24-Well Semi-Skirted PCR Plate is pre-cut into 24-well segments. Its convenient format allows for lower sample numbers. The semi skirt adds rigidity and allots space for labeling or barcoding. Available in various colors and compatible with standard multichannel pipettes. Sealing is possible with adhesive films and foils and all cap strips.
Highlights
• Conveniently pre-cut into 24-well segments
• Suitable for use in 0.2 mL thermal cycler blocks
• Semi-skirt adds rigidity and allows for labeling or barcoding
• 0.3 mL maximum well volume (when used with adhesive seals)
• Sealable using adhesive films and foils and all cap strips
Certifications
Certified free from human DNA, RNases, and DNases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Red
24
4 x 6
Virgin Polypropylene