Learn More
Thermo Scientific™ 24-Well Semi-Skirted PCR Plate
24-well semi-skirted plate for PCR and qPCR applications.
958.00 SEK - 1072.00 SEK
Specifications
Format Standard
Array 4 x 6
Material Virgin Polypropylene
No. of Wells 24
View More Specs
Products 3
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
10563795
View Documents
Thermo Scientific™
AB-0624/R
Red This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10405353
View Documents
Thermo Scientific™
AB-0624/Y
Yellow This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12657506
View Documents
Thermo Scientific™
AB-0624/W
White This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
The 24-Well Semi-Skirted PCR Plate is pre-cut into 24-well segments. Its convenient format allows for lower sample numbers. The semi skirt adds rigidity and allots space for labeling or barcoding. Available in various colors and compatible with standard multichannel pipettes. Sealing is possible with adhesive films and foils and all cap strips.
The Thermo Scientific 24-Well Semi-Skirted PCR Plate is pre-cut into 24-well segments. Its convenient format allows for lower sample numbers. The semi skirt adds rigidity and allots space for labeling or barcoding. Available in various colors and compatible with standard multichannel pipettes. Sealing is possible with adhesive films and foils and all cap strips.
Highlights
  • Conveniently pre-cut into 24-well segments
  • Suitable for use in 0.2 mL thermal cycler blocks
  • Semi-skirt adds rigidity and allows for labeling or barcoding
  • 0.3 mL maximum well volume (when used with adhesive seals)
  • Sealable using adhesive films and foils and all cap strips
Certifications
Certified free from human DNA, RNases, and DNases.
Specifications
Specifications
Standard
4 x 6
Virgin Polypropylene
24