missing translation for 'onlineSavingsMsg'
Learn More

TISAB III Buffer, Chem Lab™

Brand:  Chem Lab CL03.2007.0500

Product Code. 11925497

  • 872.00 SEK / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

0°C
Laboratory use
500 mL
65 g NH4Cl + 53 g NH4Ac + 19 g CDTA / l H2O
Soluble in water
Boiling Point: 100°C
Liquid
Buffer
Odorless
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TISAB III Buffer, Chem Lab™ > Quantity: 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.