missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM190 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24904-1-AP-20UL
This item is not returnable.
View return policy
Description
TMEM190 gene ontology annotations related to this gene include hematopoietic progenitor cell differentiation.Specifications
TMEM190 | |
Polyclonal | |
Unconjugated | |
TMEM190 | |
MDAC1, TMEM190, transmembrane protein 190 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
147744 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WZ59 | |
TMEM190 | |
TMEM190 Fusion Protein Ag19131 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |