missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF6 Mouse anti-Human, Clone: 1D1E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66498-1-IG-20UL
This item is not returnable.
View return policy
Description
Transaldolase 1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. The functional gene of transaldolase 1 is located on chromosome 11 and a pseudogene is identified on chromosome 1 but there are conflicting map locations. The second and third exon of this gene were developed by insertion of a retrotransposable element. This gene is thought to be involved in multiple sclerosis.Specifications
TRAF6 | |
Monoclonal | |
0.7 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y4K3 | |
TRAF6 | |
TRAF6 Fusion Protein Ag24696 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
1D1E1 | |
Unconjugated | |
TRAF6 | |
RING finger protein 85, RNF85, TRAF6 | |
Mouse | |
Protein G | |
RUO | |
7189 | |
-20°C | |
Liquid |