missing translation for 'onlineSavingsMsg'
Learn More

TRAF6 Mouse anti-Human, Clone: 1D1E1, Proteintech

Mouse Monoclonal Antibody

1236.20 SEK - 3313.85 SEK

Specifications

Antigen TRAF6
Clone 1D1E1
Concentration 0.7 mg/mL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16808435
View Documents
Proteintech
66498-1-IG-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16898425
View Documents
Proteintech
66498-1-IG-150UL
150 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Transaldolase 1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. The functional gene of transaldolase 1 is located on chromosome 11 and a pseudogene is identified on chromosome 1 but there are conflicting map locations. The second and third exon of this gene were developed by insertion of a retrotransposable element. This gene is thought to be involved in multiple sclerosis.
Specifications

Specifications

TRAF6
0.7 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
RING finger protein 85, RNF85, TRAF6
TRAF6
IgG1
Protein G
Antibody
1D1E1
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Mouse
Human
Q9Y4K3
7189
TRAF6 Fusion Protein Ag24696
Primary
-20°C
TRAF6
Videos
SDS
Documents

Documents

Product Certifications