Learn More
(+/-)-trans-1,2-Diaminocyclohexane, 99%, Thermo Scientific Chemicals
568.00 SEK - 5475.00 SEK
Chemical Identifiers
CAS 1121-22-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00063747
InChI Key SSJXIUAHEKJCMH-PHDIDXHHSA-N
Synonym 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
PubChem CID 43806
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine
SMILES C1CCC(C(C1)N)N
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11376926
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L06626.09
10 g
568.00 SEK
10g
11386926
View Documents
Thermo Scientific Alfa Aesar
L06626.18
50 g
1670.00 SEK
50g
11396926
View Documents
Thermo Scientific Alfa Aesar
L06626.30
250 g
5475.00 SEK
250g
Description
Description
(±)-trans-1,2-Diaminocyclohexane is employed in the synthesis of macrocyclic [3+3] hexa Schiff base. It also plays an important role in the preparation of multidentate ligands, chiral auxiliaries and chiral stationary phases. Further, it is used to prepare [2+2] macrocyclization by reacting with aliphatic dialdehydes. In addition to this, it acts as a chiral ligand, which finds application in asymmetric catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(^+)-trans-1,2-Diaminocyclohexane is employed in the synthesis of macrocyclic [3+3] hexa Schiff base. It also plays an important role in the preparation of multidentate ligands, chiral auxiliaries and chiral stationary phases. Further, it is used to prepare [2+2] macrocyclization by reacting with aliphatic dialdehydes. In addition to this, it acts as a chiral ligand, which finds application in asymmetric catalysis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
Chemical Identifiers
1121-22-8
114.192
SSJXIUAHEKJCMH-PHDIDXHHSA-N
43806
C1CCC(C(C1)N)N
C6H14N2
MFCD00063747
1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
(1R,2R)-cyclohexane-1,2-diamine
Specifications
1121-22-8
0.951
68°C (154°F)
C6H14N2
MFCD00063747
3193807
1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
SSJXIUAHEKJCMH-PHDIDXHHSA-N
(1R,2R)-cyclohexane-1,2-diamine
43806
99%
14°C to 15°C
89°C to 90°C (14 mmHg)
Amine-like
1.489
UN2735
Air sensitive; May form a precipitate
Miscible with water.
C1CCC(C(C1)N)N
114.192
114.19
(+/-)-trans-1,2-Diaminocyclohexane
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen